4-Methoxy-N-methylbenzylamine hydrochloride 95%

Supplier: Ambeed
Warning

A197594-1G A197594-250MG A197594-5G
77287-590EA 15.45 USD
77287-590 77287-592 77287-594
4-Methoxy-N-methylbenzylamine hydrochloride 95%
4-Methoxy-N-methylbenzylamine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₄ClNO
MW: 187.6700 g/mol
MDL Number: MFCD04117881
CAS Number: 876-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR