2-Methyl-1,4-phenylene bis(4-(4-(acryloyloxy)butoxy)benzoate) 97%

Supplier: Ambeed
Warning

A197500-1G A197500-25G A197500-5G
77248-774EA 22.66 USD
77248-774 77248-776 77248-778
2-Methyl-1,4-phenylene bis(4-(4-(acryloyloxy)butoxy)benzoate) 97%
2-Methyl-1,4-phenylene bis(4-(4-(acryloyloxy)butoxy)benzoate)

Precaución: for Research Use Only

Formula: C₃₅H₃₆O₁₀
MW: 616.65 g/mol
MDL Number: MFCD26143191
CAS Number: 132900-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR