2-Fluoro-6-nitrobenzaldehyde 97%

Supplier: Ambeed
Warning

A197450-10G A197450-1G A197450-250MG A197450-25G A197450-5G
77260-698EA 124 USD
77260-698 77260-700 77260-702 77260-704 77260-706
2-Fluoro-6-nitrobenzaldehyde 97%
2-Fluoro-6-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄FNO₃
MDL Number: MFCD09835320
CAS Number: 1644-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR