Acetyl-L-carnitine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (R)-3-Acetoxy-4-(trimethylammonio)butyrate Hydrochloride, (R)-2-Acetoxy-3-carboxy-N,N,N-trimethylpropan-1-aminium chloride, Acetyl-L-carnitine HCl

A197422-1000G A197422-100G A197422-25G A197422-500G A197422-5G
77274-684EA 476.89 USD
77274-684 77274-686 77274-688 77274-690 77748-249
Acetyl-L-carnitine hydrochloride 97%
Acetyl-L-carnitine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₇NO₄
MW: 239.70 g/mol
MDL Number: MFCD00082230
CAS Number: 5080-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR