4-Chloro-N-methoxy-N-methylbenzamide 97%

Supplier: Ambeed
Warning

A197269-100G A197269-10G A197269-5G
77287-584EA 119 USD
77287-584 77287-586 77287-588
4-Chloro-N-methoxy-N-methylbenzamide 97%
4-Chloro-N-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₂
MDL Number: MFCD02684308
CAS Number: 122334-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR