1-Chloro-2,4-dimethyl-5-nitrobenzene 95%

Supplier: Ambeed
Warning

A197195-1G A197195-250MG
77237-068EA 127.72 USD
77237-068 77237-070
1-Chloro-2,4-dimethyl-5-nitrobenzene 95%
1-Chloro-2,4-dimethyl-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MW: 185.608 g/mol
MDL Number: MFCD00547853
CAS Number: 69383-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR