WAY-100635 xMaleate 99+%

Supplier: Ambeed
Warning

Synonyms: N-(2-(4-(2-Methoxyphenyl)piperazin-1-yl)ethyl)-N-(pyridin-2-yl)cyclohexanecarboxamide xmaleate, WAY-100635 Maleate, 5-HT1A Serotonis Receptor Antagonist

A197128-10MG A197128-1MG A197128-5MG
77237-062EA 30 USD
77237-062 77237-064 77237-066
WAY-100635 xMaleate 99+%
WAY-100635 xMaleate

Precaución: for Research Use Only

Formula: C₂₉H₃₈N₄O₆
MW: 538.64 g/mol
MDL Number: MFCD01321056
CAS Number: 634908-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR