2-Cyano-N-(2,4-dichloro-5-methoxyphenyl)acetamide 97%

Supplier: Ambeed
Warning

A196994-10G A196994-1G
77260-694EA 57.68 USD
77260-694 77260-696
2-Cyano-N-(2,4-dichloro-5-methoxyphenyl)acetamide 97%
2-Cyano-N-(2,4-dichloro-5-methoxyphenyl)acetamide

Precaución: for Research Use Only

Formula: C₁₀H₈Cl₂N₂O₂
MW: 259.09 g/mol
MDL Number: MFCD08143656
CAS Number: 846023-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR