Fmoc-N-Me-D-Trp(Boc)-OH 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-N-Me-Trp(Boc)-OH

A196922-100MG A196922-1G A196922-250MG
77237-056EA 53 USD
77237-056 77237-058 77237-060
Fmoc-N-Me-D-Trp(Boc)-OH 97%
Fmoc-N-Me-D-Trp(Boc)-OH

Precaución: for Research Use Only

Formula: C₃₂H₃₂N₂O₆
MW: 540.60 g/mol
MDL Number: MFCD30475981
CAS Number: 2044709-90-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR