2-(Methylamino)nicotinonitrile 98%

Supplier: Ambeed
Warning

A196836-1G A196836-250MG A196836-5G
77248-746EA 39.14 USD
77248-746 77248-748 77248-750
2-(Methylamino)nicotinonitrile 98%
2-(Methylamino)nicotinonitrile

Precaución: for Research Use Only

Formula: C₇H₇N₃
MW: 133.1506 g/mol
MDL Number: MFCD08689752
CAS Number: 52583-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR