2',7'-Dichlorofluorescein 3',6'-diacetate 98%

Supplier: Ambeed
Warning

Synonyms: 2',7'-Dichlorofluorescin diacetate, 2,7-Diazaspiro[4.5]decane-7-carboxylic acid tert-butyl ester, 2',7'-Dichloro-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-diyl diacetate, H₂DCFDA

A196793-1G A196793-250MG A196793-5G
77248-740EA 36 USD
77248-740 77248-742 77248-744
2',7'-Dichlorofluorescein 3',6'-diacetate 98%
2',7'-Dichlorofluorescein 3',6'-diacetate

Precaución: for Research Use Only

Formula: C₂₄H₁₄Cl₂O₇
MDL Number: MFCD00037501
CAS Number: 2044-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR