4-Chloro-2-methylpyrimidine 96%

Supplier: Ambeed
Warning

A196750-100G A196750-10G A196750-1G A196750-25G A196750-5G
77287-568EA 342.99 USD
77287-568 77287-570 77287-572 77287-574 77287-576
4-Chloro-2-methylpyrimidine 96%
4-Chloro-2-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₅ClN₂
MDL Number: MFCD00234069
CAS Number: 4994-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR