2-Amino-6-chloro-5-methylpyridine 97%

Supplier: Ambeed
Warning

A196704-10G A196704-1G A196704-250MG A196704-5G
77274-648EA 45 USD
77274-648 77274-650 77274-652 77274-654
2-Amino-6-chloro-5-methylpyridine 97%
2-Amino-6-chloro-5-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂
MDL Number: MFCD13193513
CAS Number: 442129-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR