2,4-Dihydroxy-3-nitro-6-picoline 98%

Supplier: Ambeed
Warning

Synonyms: 2,4-Dihydroxy-6-methyl-3-nitropyridine, 4-Hydroxy-6-methyl-3-nitropyridin-2(1H)-one, 4-Hydroxy-6-methyl-3-nitro-2-pyridone

A196698-100G A196698-10G A196698-1G A196698-25G A196698-5G
77287-558EA 148 USD
77287-558 77287-560 77287-562 77287-564 77287-566
2,4-Dihydroxy-3-nitro-6-picoline 98%
2,4-Dihydroxy-3-nitro-6-picoline

Precaución: for Research Use Only

Formula: C₆H₆N₂O₄
MW: 170.12 g/mol
MDL Number: MFCD00167612
CAS Number: 4966-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR