(S)-1-{(S)-2-[Bis(4-methoxy-3,5-dimethylphenyl)phosphino]ferrocenyl}-ethylbis(2-methylphenyl)phosphine 98%

Supplier: Ambeed
Warning

A196671-100MG A196671-250MG A196671-1G
77248-734EA 87.55 USD
77248-734 77248-736 77746-242
(S)-1-{(S)-2-[Bis(4-methoxy-3,5-dimethylphenyl)phosphino]ferrocenyl}-ethylbis(2-methylphenyl)phosphine 98%
(S)-1-{(S)-2-[Bis(4-methoxy-3,5-dimethylphenyl)phosphino]ferrocenyl}-ethylbis(2-methylphenyl)phosphine

Precaución: for Research Use Only

Formula: C₄₄H₄₈FeO₂P₂
MW: 726.64 g/mol
MDL Number: MFCD08561167
CAS Number: 849924-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR