2,4-Dimethylbenzaldehyde 95%

Supplier: Ambeed
Warning

A196619-1G A196619-25G A196619-5G
77248-728EA 7.21 USD
77248-728 77248-730 77248-732
2,4-Dimethylbenzaldehyde 95%
2,4-Dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O
MW: 134.18 g/mol
MDL Number: MFCD00003340
CAS Number: 15764-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR