Diethyl naphthalene-2,6-dicarboxylate 97%

Supplier: Ambeed
Warning

A196599-250MG A196599-1G
77237-054EA 30.9 USD
77237-054 77746-241
Diethyl naphthalene-2,6-dicarboxylate 97%
Diethyl naphthalene-2,6-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₄
MW: 272.3 g/mol
MDL Number: MFCD30475487
CAS Number: 15442-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR