Dinaciclib 98%

Supplier: Ambeed
Warning

A196584-10MG A196584-1MG A196584-5MG A196584-50MG
77237-048EA 106 USD
77237-048 77237-050 77237-052 77746-240
Dinaciclib 98%
Dinaciclib

Precaución: for Research Use Only

Formula: C₂₁H₂₈N₆O₂
MDL Number: MFCD16037702
CAS Number: 779353-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR