2-(Benzimidazolylthio)acetic acid hydrazide 95%

Supplier: Ambeed
Warning

A196524-1G A196524-25G A196524-5G
77248-722EA 18 USD
77248-722 77248-724 77248-726
2-(Benzimidazolylthio)acetic acid hydrazide 95%
2-(Benzimidazolylthio)acetic acid hydrazide

Precaución: for Research Use Only

Formula: C₉H₁₀N₄O₄S
MDL Number: MFCD00171027
CAS Number: 30065-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR