tert-Butyl 6-bromo-1-oxo-3,4-dihydroisoquinoline-2(1H)-carboxylate 95%

Supplier: Ambeed
Warning

A196255-100MG A196255-250MG A196255-1G
77260-688EA 23.69 USD
77260-688 77260-690 77593-270
tert-Butyl 6-bromo-1-oxo-3,4-dihydroisoquinoline-2(1H)-carboxylate 95%
tert-Butyl 6-bromo-1-oxo-3,4-dihydroisoquinoline-2(1H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₆BrNO₃
MW: 326.18 g/mol
MDL Number: MFCD28399182
CAS Number: 1214900-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR