3-Fluoro-2-methoxybenzeneboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-(3-Fluoro-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Fluoro-2-methoxyphenylboronic acid pinacol ester

A196195-100MG A196195-1G A196195-250MG
77260-682EA 14.42 USD
77260-682 77260-684 77260-686
3-Fluoro-2-methoxybenzeneboronic acid pinacol ester 98%
3-Fluoro-2-methoxybenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BFO₃
MW: 252.09 g/mol
MDL Number: MFCD16996297
CAS Number: 1417301-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR