2-Pentylthiophene 97%

Supplier: Ambeed
Warning

A196154-100G A196154-25G A196154-5G
77248-700EA 91 USD
77248-700 77248-702 77248-704
2-Pentylthiophene 97%
2-Pentylthiophene

Precaución: for Research Use Only

Formula: C₉H₁₄S
MDL Number: MFCD00041017
CAS Number: 4861-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR