2,4,5-Trifluoro-3-ethoxybenzoic acid 97%

Supplier: Ambeed
Warning

A195613-100MG A195613-1G
77248-694EA 28.84 USD
77248-694 77248-696
2,4,5-Trifluoro-3-ethoxybenzoic acid 97%
2,4,5-Trifluoro-3-ethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₇F₃O₃
MW: 220.145 g/mol
MDL Number: MFCD08459286
CAS Number: 169507-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR