3,5-Dimethoxyphenylboronic acid pinacol ester 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(3,5-Dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3-Dimethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 3,5-Dimethoxyphenylboronic acid Pinacol ester
A195509-100G
A195509-10G
A195509-1G
A195509-250MG
A195509-25G
A195509-5G
77237-022EA
897.13
USD
77237-022
77237-024
77237-026
77237-028
77237-030
77237-032
3,5-Dimethoxyphenylboronic acid pinacol ester 95%
3,5-Dimethoxyphenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₄H₂₁BO₄ MW: 264.13 g/mol |
MDL Number:
MFCD05865191 CAS Number: 365564-07-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...