(1R,2R)-N,N'-Bis(2-acetyl-3-oxo-2-butenylidene)-1,2-dimesitylethylenediaminato cobalt(II) 98%

Supplier: Ambeed
Warning

Synonyms: (R)-AMAC

A195380-100MG A195380-250MG
77248-686EA 121 USD
77248-686 77248-688
(1R,2R)-N,N'-Bis(2-acetyl-3-oxo-2-butenylidene)-1,2-dimesitylethylenediaminato cobalt(II) 98%
(1R,2R)-N,N'-Bis(2-acetyl-3-oxo-2-butenylidene)-1,2-dimesitylethylenediaminato cobalt(II)

Precaución: for Research Use Only

Formula: C₃₂H₃₈CoN₂O₄
MW: 573.60 g/mol
MDL Number: MFCD06797061
CAS Number: 361346-80-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR