Ethyl 5-bromo-6-chloro-1H-indole-2-carboxylate 95%

Supplier: Ambeed
Warning

A195298-100MG A195298-1G A195298-250MG
77260-640EA 49.44 USD
77260-640 77260-642 77260-644
Ethyl 5-bromo-6-chloro-1H-indole-2-carboxylate 95%
Ethyl 5-bromo-6-chloro-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₉BrClNO₂
MW: 302.55 g/mol
MDL Number: MFCD21363140
CAS Number: 1923237-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR