4-(Hydroxymethyl)-1,3-oxazole 95%

Supplier: Ambeed
Warning

Synonyms: (1,3-Oxazol-4-yl)methanol, 4-(Hydroxymethyl)oxazole

A195231-250MG A195231-1G A195231-100MG
77287-538EA 36.05 USD
77287-538 77593-258 77748-241
4-(Hydroxymethyl)-1,3-oxazole 95%
4-(Hydroxymethyl)-1,3-oxazole

Precaución: for Research Use Only

Formula: C₄H₅NO₂
MW: 99.088 g/mol
MDL Number: MFCD08703646
CAS Number: 155742-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR