6-Hydroxy-8-methyl-8-azabicyclo[3.2.1]octan-3-yl 3-hydroxy-2-phenylpropanoate 97%

Supplier: Ambeed
Warning

A195229-100G A195229-10G A195229-1G A195229-250MG A195229-25G A195229-5G
77248-668EA 1343.12 USD
77248-668 77248-670 77248-672 77248-674 77248-676 77248-678
6-Hydroxy-8-methyl-8-azabicyclo[3.2.1]octan-3-yl 3-hydroxy-2-phenylpropanoate 97%
6-Hydroxy-8-methyl-8-azabicyclo[3.2.1]octan-3-yl 3-hydroxy-2-phenylpropanoate

Precaución: for Research Use Only

Formula: C₁₇H₂₃NO₄
MW: 305.36 g/mol
MDL Number: MFCD01732987
CAS Number: 17659-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR