4-(Dimethylamino)-1-butanol 98%

Supplier: Ambeed
Danger

A195186-5G A195186-25G
77287-536EA 35 USD
77287-536 77748-240
4-(Dimethylamino)-1-butanol 98%
4-(Dimethylamino)-1-butanol

Precaución: for Research Use Only

Formula: C₆H₁₅NO
MDL Number: MFCD00020624
CAS Number: 13330-96-6
UN: 2735
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR