4,6-Dibromobenzo[d]thiazol-2-amine 98%

Supplier: Ambeed
Warning

A195107-100MG A195107-1G A195107-250MG A195107-5G
77248-652EA 6 USD
77248-652 77248-654 77248-656 77248-658
4,6-Dibromobenzo[d]thiazol-2-amine 98%
4,6-Dibromobenzo[d]thiazol-2-amine

Precaución: for Research Use Only

Formula: C₇H₄Br₂N₂S
MW: 307.99 g/mol
MDL Number: MFCD02031522
CAS Number: 16582-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR