Methyl 2-(6-bromo-1H-indol-3-yl)acetate 97%

Supplier: Ambeed
Warning

A195093-100MG A195093-1G A195093-250MG
77248-646EA 126.69 USD
77248-646 77248-648 77248-650
Methyl 2-(6-bromo-1H-indol-3-yl)acetate 97%
Methyl 2-(6-bromo-1H-indol-3-yl)acetate

Precaución: for Research Use Only

Formula: C₁₁H₁₀BrNO₂
MW: 268.11 g/mol
MDL Number: MFCD18452338
CAS Number: 152213-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR