tert-Butyl 4-formyl-5-methoxy-7-methyl-1H-indole-1-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A195090-100MG
A195090-250MG
A195090-1G
77260-626EA
33.99
USD
77260-626
77260-628
77748-238
tert-Butyl 4-formyl-5-methoxy-7-methyl-1H-indole-1-carboxylate 97%
tert-Butyl 4-formyl-5-methoxy-7-methyl-1H-indole-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₆H₁₉NO₄ MW: 289.32 g/mol |
MDL Number:
MFCD30609524 CAS Number: 1481631-51-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...