1-((2-((2-Chloropyridin-4-yl)amino)-4'-(cyclopropylmethyl)-[4,5'-bipyrimidin]-2'-yl)amino)-2-methylpropan-2-ol ≥98%

Supplier: Ambeed
Warning

Synonyms: Vps34-IN-1

A194997-10MG A194997-1MG A194997-50MG A194997-5MG A194997-100MG A194997-250MG
77237-014EA 42.23 USD
77237-014 77237-016 77237-018 77237-020 77600-582 77615-646
1-((2-((2-Chloropyridin-4-yl)amino)-4'-(cyclopropylmethyl)-[4,5'-bipyrimidin]-2'-yl)amino)-2-methylpropan-2-ol ≥98%
1-((2-((2-Chloropyridin-4-yl)amino)-4'-(cyclopropylmethyl)-[4,5'-bipyrimidin]-2'-yl)amino)-2-methylpropan-2-ol

Precaución: for Research Use Only

Formula: C₂₁H₂₄ClN₇O
MW: 425.91 g/mol
MDL Number: MFCD28963905
CAS Number: 1383716-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR