3-(4-Chlorobenzylidene)-1-(4-hydroxyphenyl)-5-phenyl-1,3-dihydro-2H-pyrrol-2-one 98%

Supplier: Ambeed
Warning

A194870-1MG A194870-5MG
77274-614EA 221.45 USD
77274-614 77274-616 77274-618 77274-620
3-(4-Chlorobenzylidene)-1-(4-hydroxyphenyl)-5-phenyl-1,3-dihydro-2H-pyrrol-2-one 98%
3-(4-Chlorobenzylidene)-1-(4-hydroxyphenyl)-5-phenyl-1,3-dihydro-2H-pyrrol-2-one

Precaución: for Research Use Only

Formula: C₂₃H₁₆ClNO₂
MDL Number: MFCD01534890
CAS Number: 328998-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR