2,5-Diaminoanisole 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxybenzene-1,4-diamine

A194765-100MG A194765-1G A194765-250MG A194765-5G
77260-608EA 65.92 USD
77260-608 77260-610 77260-612 77260-614
2,5-Diaminoanisole 97%
2,5-Diaminoanisole

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O
MW: 138.17 g/mol
MDL Number: MFCD00466319
CAS Number: 5307-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR