Ethyl 3-(2,4-dichlorophenyl)-3-oxopropanoate 95%

Supplier: Ambeed
Warning

A194764-10G A194764-1G A194764-250MG A194764-5G
77274-608EA 159.65 USD
77274-608 77274-610 77274-612 77748-236
Ethyl 3-(2,4-dichlorophenyl)-3-oxopropanoate 95%
Ethyl 3-(2,4-dichlorophenyl)-3-oxopropanoate

Precaución: for Research Use Only

Formula: C₁₁H₁₀Cl₂O₃
MW: 261.10 g/mol
MDL Number: MFCD03424829
CAS Number: 60868-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR