Methyl-2-amino-1,3-benzothiazole-6-carboxylate 98%

Supplier: Ambeed
Warning

A194598-10G A194598-250MG A194598-1G
77260-604EA 46 USD
77260-604 77260-606 77746-236
Methyl-2-amino-1,3-benzothiazole-6-carboxylate 98%
Methyl-2-amino-1,3-benzothiazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂S
MDL Number: MFCD00468672
CAS Number: 66947-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR