Benzo[b]thiophene-3-carboxaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: Benzo[b]thiophene-3-carbaldehyde

A194549-100G A194549-1G A194549-250MG A194549-25G A194549-5G
77260-594EA 455.26 USD
77260-594 77260-596 77260-598 77260-600 77260-602
Benzo[b]thiophene-3-carboxaldehyde 97%
Benzo[b]thiophene-3-carboxaldehyde

Precaución: for Research Use Only

Formula: C₉H₆OS
MW: 162.2083 g/mol
MDL Number: MFCD00052376
CAS Number: 5381-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR