Homogentisic acid-γ-lactone 98%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dihydro-5-hydroxybenzofuran-2-one, 2,5-Dihydroxyphenylacetic Acid γ-Lactone, 5-Hydroxybenzofuran-2(3H)-one, 5-Hydroxy-2-coumaranone

A194533-1G A194533-250MG A194533-5G
77237-002EA 50.47 USD
77237-002 77237-004 77237-006
Homogentisic acid-γ-lactone 98%
Homogentisic acid-gamma-lactone

Precaución: for Research Use Only

Formula: C₈H₆O₃
MW: 150.13 g/mol
MDL Number: MFCD00005857
CAS Number: 2688-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR