(S)-4-(Pyrrolidin-3-yl)morpholine dihydrochloride 97%

Supplier: Ambeed
Warning

A194419-100MG A194419-250MG
77236-998EA 88.58 USD
77236-998 77237-000
(S)-4-(Pyrrolidin-3-yl)morpholine dihydrochloride 97%
(S)-4-(Pyrrolidin-3-yl)morpholine dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₈Cl₂N₂O
MW: 229.14 g/mol
MDL Number: MFCD22741807
CAS Number: 2829292-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR