4-Methylbenzimidazole 98%

Supplier: Ambeed
Warning

A194340-250MG A194340-5G A194340-1G
77287-532EA 7 USD
77287-532 77567-820 77748-235
4-Methylbenzimidazole 98%
4-Methylbenzimidazole

Precaución: For research use only.

Formula: C₈H₈N₂
MDL Number: MFCD09842603
CAS Number: 4887-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR