3'-Hydroxypropiophenone 97%

Supplier: Ambeed
Warning

A194186-1G A194186-250MG A194186-5G A194186-100G
77248-632EA 6 USD
77248-632 77248-634 77248-636 77746-234
3'-Hydroxypropiophenone 97%
3'-Hydroxypropiophenone

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MDL Number: MFCD03428514
CAS Number: 13103-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR