N-Methylpiperidin-3-amine dihydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 3-(Methylamino)piperidine dihydrochloride

A194146-10G A194146-25G A194146-5G
77287-526EA 21.63 USD
77287-526 77287-528 77287-530
N-Methylpiperidin-3-amine dihydrochloride 95%
N-Methylpiperidin-3-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₆Cl₂N₂
MW: 187.1100 g/mol
MDL Number: MFCD00673143
CAS Number: 127294-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR