5-tert-Butyl-1-ethyl 3-aminopyrrolo[3,4-c]pyrazole-1,5(4H,6H)-dicarboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-tert-Butyl 1-ethyl 3-aminopyrrolo[3,4-c]pyrazole-1,5(4H,6H)-dicarboxylate
A193972-1G
A193972-250MG
77236-984EA
92.7
USD
77236-984
77236-986
5-tert-Butyl-1-ethyl 3-aminopyrrolo[3,4-c]pyrazole-1,5(4H,6H)-dicarboxylate 95%
5-tert-Butyl-1-ethyl 3-aminopyrrolo[3,4-c]pyrazole-1,5(4H,6H)-dicarboxylate
Precaución: for Research Use Only
Formula:
C13H20N4O4 MW: 296.3223 g/mol |
MDL Number:
MFCD08061258 CAS Number: 398495-65-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...