(Hexamethylbenzene)ruthenium(II) dichloride 98%

Supplier: Ambeed
Warning

Synonyms: Dichloro(hexamethylbenzene)ruthenium(II) Dimer, (Hexamethylbenzene)ruthenium(II) dichloride dimer

A193969-100MG A193969-1G A193969-250MG
77248-620EA 65 USD
77248-620 77248-622 77248-624
(Hexamethylbenzene)ruthenium(II) dichloride 98%
(Hexamethylbenzene)ruthenium(II) dichloride

Precaución: for Research Use Only

Formula: C₂₄H₃₆Cl₄Ru₂
MW: 668.49 g/mol
MDL Number: MFCD03701102
CAS Number: 67421-02-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR