trans-2-Methoxycinnamic acid 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
(E)-3-(2-Methoxyphenyl)acrylic acid, trans-O-Methyl-o-coumaric acid, (E)-3-(2-Methoxyphenyl)-2-propenoic acid
A193678-100G
A193678-25G
A193678-5G
77260-570EA
107.12
USD
77260-570
77260-572
77260-574
trans-2-Methoxycinnamic acid 99%
trans-2-Methoxycinnamic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₀O₃ MW: 178.19 g/mol |
MDL Number:
MFCD00064238 CAS Number: 1011-54-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...