Cyclo(L-arginylglycyl-L-α-aspartyl-D-tyrosyl-L-lysyl) di2,2,2-Trifluoroacetic acid salt 98%

Supplier: Ambeed
Danger

Synonyms: 2,2,2-Trifluoroacetic acid compound with 2-((2S,5R,8S,11S)-8-(4-aminobutyl)-11-(3-guanidinopropyl)-5-(4-hydroxybenzyl)-3,6,9,12,15-pentaoxo-1,4,7,10,13-pentaazacyclopentadecan-2-yl)acetic acid (2:1)

A193345-10MG A193345-1MG A193345-25MG A193345-50MG A193345-5MG
77260-546EA 442.9 USD
77260-546 77260-548 77260-550 77260-552 77260-554 77260-556
Cyclo(L-arginylglycyl-L-α-aspartyl-D-tyrosyl-L-lysyl) di2,2,2-Trifluoroacetic acid salt 98%
Cyclo(L-arginylglycyl-L-α-aspartyl-D-tyrosyl-L-lysyl) di2,2,2-Trifluoroacetic acid salt

Precaución: for Research Use Only

Formula: C₃₁H₄₃F₆N₉O₁₂
MW: 847.72 g/mol
MDL Number: MFCD20134101
CAS Number: 250612-42-1
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR