1-Butanesulphonic acid sodium salt 98%

Supplier: Ambeed
Warning

Synonyms: Sodium 1-butanesulphonate, Sodium butane-1-sulphonate, Sodium 1-butansulphonate, Butane-1-sulphonic acid sodium salt

A193325-100G A193325-10G A193325-5G
77236-976EA 195 USD
77236-976 77236-978 77236-980
1-Butanesulphonic acid sodium salt 98%
1-Butanesulphonic acid sodium salt

Precaución: for Research Use Only

Formula: C₄H₉NaO₃S
MDL Number: MFCD00007540
CAS Number: 2386-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR