7-Methoxy-1H-pyrrolo[2,3-c]pyridine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
7-Methoxy-6-azaindole
A193318-100MG
A193318-1G
A193318-250MG
77236-970EA
32.96
USD
77236-970
77236-972
77236-974
7-Methoxy-1H-pyrrolo[2,3-c]pyridine 98%
7-Methoxy-1H-pyrrolo[2,3-c]pyridine
Precaución: for Research Use Only
Formula:
C₈H₈N₂O MW: 148.17 g/mol |
MDL Number:
MFCD09864857 CAS Number: 160590-40-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...