2-Nitro-4-(trifluoromethyl)anisole 98%

Supplier: Ambeed
Danger

Synonyms: 4-Methoxy-3-nitrobenzotrifluoride, 1-Methoxy-2-nitro-4-(trifluoromethyl)benzene

A193305-10G A193305-25G A193305-5G
77236-964EA 9.27 USD
77236-964 77236-966 77236-968
2-Nitro-4-(trifluoromethyl)anisole 98%
2-Nitro-4-(trifluoromethyl)anisole

Precaución: for Research Use Only

Formula: C₈H₆F₃NO₃
MW: 221.14 g/mol
MDL Number: MFCD00007099
CAS Number: 394-25-2
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR